Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cys-TAT(47-57) TFA | 98.1% | 1661.99 | 10 MG
SDP

Catalog No. 5000423930
Encompass_Preferred

Cys-TAT(47-57) (Cys-[HIV-Tat (47-57)]) is an arginine rich cell penetrating peptide derived from the HIV-1 transactivating protein. This product is intended for research use only and is not sold to patients.

  • Purity: 98.1%
  • Molecular weight: 1661.99 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in H2O: 100 mg/mL (ultrasonic needed)
  • Solubility in DMSO: ≥ 100 mg/mL

Catalog No. 50-004-23930 Supplier Medchemexpress LLC Supplier No. HYP1801A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cys-TAT(47-57) (Cys-[HIV-Tat (47-57)]) is an arginine rich cell penetrating peptide derived from the HIV-1 transactivating protein. This product is intended for research use only and is not sold to patients.

  • Purity: 98.1%
  • Molecular weight: 1661.99 (free base)
  • Appearance: Solid
  • Color: White to off-white
  • Solubility in H2O: 100 mg/mL (ultrasonic needed)
  • Solubility in DMSO: ≥ 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.