Learn More
CPC Scientific H-Thr-Tyr-Ile-Cys(benzyl)-Glu-Val-Glu-Asp-Gln-Lys-Glu-Glu-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific HIV001A
Encompass
CD4 fragment that inhibits CD4 cell infection by HIV and HIV-induced cell fusion. As a cardioaccelerating peptide that has been derived from the corpus cardiacum of the American cockroach, Periplaneta Americana, corazonin is the most potent insect cardioactive neuropeptide.ONE-LETTER SEQUENCE: TYI-Cys(benzyl)-EVEDQKEEMOLECULAR FORMULA: C69H102N14O26S1MOLECULAR WEIGHT:1575.69STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [123380-68-7], SYNONYMS: [Cys(Bzl)]84-CD4 Fragment 81-92RESEARCH AREA: AntimicrobialREFERENCES: P.L. Nara et al., PNAS., 86, 9159 (1989)
Catalog No. 50-210-9533