Learn More
Cayman Chemical CysLT1 Receptor Polyclonal Antibody, 1 ea
SDP
Catalog No. 502954915
Encompass_Preferred
Immunogen: Synthetic peptide from the C-termianl region of human CysLT1 • Host: Rabbit • Species Reactivity: (+) Human and murine • Application: WB • The CysLT1 receptor binds LTD4, LTC4, and LTE4, playing a major role in asthma by facillitating airway and pulmonary vascular smooth muscle contraction, increased vascular permeability, and stimulating mucus secretion. The mRNA for the human CysLT1 receptor is expressed in spleen and peripheral blood leukocytes with smaller amounts in lung, placenta, and small intestine.
Catalog No. 50-295-4915 Supplier Cayman Chemical Supplier No. 1205001 EA
Immunogen: Synthetic peptide from the C-termianl region of human CysLT1 • Host: Rabbit • Species Reactivity: (+) Human and murine • Application: WB • The CysLT1 receptor binds LTD4, LTC4, and LTE4, playing a major role in asthma by facillitating airway and pulmonary vascular smooth muscle contraction, increased vascular permeability, and stimulating mucus secretion. The mRNA for the human CysLT1 receptor is expressed in spleen and peripheral blood leukocytes with smaller amounts in lung, placenta, and small intestine.