Learn More
Cayman Chemical CysLT1 RECPTR PolycLNal FI 1ea
SDP
Catalog No. 502972668
Encompass_Preferred
Immunogen: Synthetic peptide from the C-terminal region of CysLT1 receptor • Host: Rabbit • Species Reactivity: (+) Human and mouse CysLT1 receptor • Application(s): FC • The CysLT1 receptor binds LTD4, LTC4, and LTE4, playing a major role in asthma by facillitating airway and pulmonary vascular smooth muscle contraction, increased vascular permeability, and stimulating mucus secretion. The mRNA for the human CysLT1 receptor is expressed in spleen and peripheral blood leukocytes with smaller amounts in lung, placenta, and small intestine.
Catalog No. 50-297-2668 Supplier Cayman Chemical Supplier No. 147131 EA
Immunogen: Synthetic peptide from the C-terminal region of CysLT1 receptor • Host: Rabbit • Species Reactivity: (+) Human and mouse CysLT1 receptor • Application(s): FC • The CysLT1 receptor binds LTD4, LTC4, and LTE4, playing a major role in asthma by facillitating airway and pulmonary vascular smooth muscle contraction, increased vascular permeability, and stimulating mucus secretion. The mRNA for the human CysLT1 receptor is expressed in spleen and peripheral blood leukocytes with smaller amounts in lung, placenta, and small intestine.