Learn More
Cayman Chemical CysLT2 ReceptrC-Term Poly 1ea
SDP
Catalog No. 502955449
Encompass_Preferred
Immunogen: Synthetic peptide from the C-terminal region of human CysLT2 Receptor • Host: Rabbit • Cross Reactivity: (+) CysLT1 • Cross Reactivity: (+) Human; (-) Rat • Application(s): ELISA, FC, IHC, and WB • The CysLT2 receptor binds LTD4, LTC4, and LTE4 playing a major role in asthma by facillitating airway and pulmonary vascular smooth muscle contraction, increased vascular permeability, and stimulating mucus secretion. The mRNA for the human CysLT2 receptor is expressed in lung macrophages, airway smooth muscle, cardiac Purkinje cells, adrenal medulla cells, peripheral blood leukocytes, spleen, placenta, and brain.
Catalog No. 50-295-5449 Supplier Cayman Chemical Supplier No. 1205501 EA
Immunogen: Synthetic peptide from the C-terminal region of human CysLT2 Receptor • Host: Rabbit • Cross Reactivity: (+) CysLT1 • Cross Reactivity: (+) Human; (-) Rat • Application(s): ELISA, FC, IHC, and WB • The CysLT2 receptor binds LTD4, LTC4, and LTE4 playing a major role in asthma by facillitating airway and pulmonary vascular smooth muscle contraction, increased vascular permeability, and stimulating mucus secretion. The mRNA for the human CysLT2 receptor is expressed in lung macrophages, airway smooth muscle, cardiac Purkinje cells, adrenal medulla cells, peripheral blood leukocytes, spleen, placenta, and brain.