Learn More
Cayman Chemical CysLT2 ReceptrN-Term Bloc 1ea
SDP
Catalog No. 502946219
Encompass_Preferred
A 5-HT1D receptor antagonist (Ki 0.013 UM in CHO cell membranes expressing the human receptor) selective for 5-HT1D over several other G protein-coupled 5-HT receptor subtypes (Kis 0.25-6.3 UM) but does bind 5-HT1A and 5-HT2B receptors (Kis 0.02 and 0.04 UM respectively also acts as a partial agonist at 5-HT1D receptors (EC50 0.008 UM in a [35S]GTPYS binding assay)
Catalog No. 50-294-6219 Supplier Cayman Chemical Supplier No. 3205601 EA
A 5-HT1D receptor antagonist (Ki 0.013 UM in CHO cell membranes expressing the human receptor) selective for 5-HT1D over several other G protein-coupled 5-HT receptor subtypes (Kis 0.25-6.3 UM) but does bind 5-HT1A and 5-HT2B receptors (Kis 0.02 and 0.04 UM respectively also acts as a partial agonist at 5-HT1D receptors (EC50 0.008 UM in a [35S]GTPYS binding assay)