Learn More
Molecular Dimensions CYSTALIZATION SCREEN 96 X 10ML
SDP
Catalog No. NC0269752
Encompass_Preferred
Protein Crystallography. ProPlex screen (96 x 10mL) is formulated for the crystallization of Protein comPlexes. Based on results of Protein Complex Crystallization Database. Satisfies dual requirements: -Maintain protein-protein interactions, -Reduce solubility of complex.
Catalog No. NC0269752 Supplier Molecular Dimensions Supplier No. MD138
Protein Crystallography. ProPlex screen (96 x 10mL) is formulated for the crystallization of Protein comPlexes. Based on results of Protein Complex Crystallization Database. Satisfies dual requirements: -Maintain protein-protein interactions, -Reduce solubility of complex.