Learn More
AVANTOR MASTERFLEX (VWR LLC) Luer fitting, male luer lock x 1/4"" hose barb, Yellow, 25/pk
Catalog No. NC1496720
Encompass
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A.
Catalog No. NC1496720 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4552026
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A.