Learn More
Cayman Chemical CystamIn-d8hydrochlorIde 10mg
SDP
Catalog No. 502934038
Encompass_Preferred
An internal standard for the quantification of cystamine by GC- or LC-MS
Catalog No. 50-293-4038 Supplier Cayman Chemical Supplier No. 2677510 MG
An internal standard for the quantification of cystamine by GC- or LC-MS