Learn More
TARGETMOL CHEMICALS INC Cystamine 500MG
SDP
Catalog No. 502880035
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cystamine (2,2'-Disulfanediyldiethanamine) is an inhibitor of transglutaminase and inhibits caspase-3 with an IC50 value of 23.6 uM. Cystamine can be used in studies about Huntington's diseases. Purity 99.41%
Catalog No. 50-288-0035 Supplier TARGETMOL CHEMICALS INC Supplier No. T15035500MG
Also available in 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cystamine (2,2'-Disulfanediyldiethanamine) is an inhibitor of transglutaminase and inhibits caspase-3 with an IC50 value of 23.6 uM. Cystamine can be used in studies about Huntington's diseases. Purity 99.41%