Learn More
Medchemexpress LLC Cystamine | 51-85-4 | 96.6% | 152.28 g/mol | C4H12N2S2 | 50MG
SDP
Catalog No. 5000236254
Encompass_Preferred
Cystamine is the disulfide form of cysteamine and functions as an orally active transglutaminase (Tgase) inhibitor used in biochemical and neuroprotection research. The compound is supplied for research use and is offered in multiple pack sizes and solution formulations to support in vitro assays and in vivo studies.
  • Acts as a transglutaminase inhibitor.
  • Available in multiple pack sizes and solution formulations.
  • Suitable for biochemical assays and neuroprotection research.
  • Molecular weight 152.28 g/mol and formula C4H12N2S2.
  • Reported purity approximately 96.6% (verify with supplier documentation).
Catalog No. 50-002-36254 Supplier Medchemexpress LLC Supplier No. HY12447650MG
Cystamine is the disulfide form of cysteamine and functions as an orally active transglutaminase (Tgase) inhibitor used in biochemical and neuroprotection research. The compound is supplied for research use and is offered in multiple pack sizes and solution formulations to support in vitro assays and in vivo studies.
  • Acts as a transglutaminase inhibitor.
  • Available in multiple pack sizes and solution formulations.
  • Suitable for biochemical assays and neuroprotection research.
  • Molecular weight 152.28 g/mol and formula C4H12N2S2.
  • Reported purity approximately 96.6% (verify with supplier documentation).