Learn More
Medchemexpress LLC Cystamine | 51-85-4 | MFCD01002952 | 152.28 g/mol | C4H12N2S2 | 10MM 1ML
SDP
Catalog No. 5000238912
Encompass_Preferred
Cystamine is the disulfide form of cysteamine supplied as a 10 mM solution in DMSO (1 mL) for biochemical and cell-based assays. It acts as a transglutaminase (Tgase) inhibitor and shows caspase-3 inhibition, commonly used in apoptosis and signaling pathway studies. CAS 51-85-4; formula C4H12N2S2; MW 152.28 g/mol.
  • Supplied as a ready-to-use 10 mM stock solution in DMSO.
  • Convenient 1 mL aliquot for small-scale assays.
  • Inhibits transglutaminase and caspase-3 in biochemical assays.
  • Recommended storage: pure form -20°C; in solvent -80°C or -20°C.
  • Datasheet includes preparation guidance for stock solutions.
Catalog No. 50-002-38912 Supplier Medchemexpress LLC Supplier No. HY12447610MM 1ML
Cystamine is the disulfide form of cysteamine supplied as a 10 mM solution in DMSO (1 mL) for biochemical and cell-based assays. It acts as a transglutaminase (Tgase) inhibitor and shows caspase-3 inhibition, commonly used in apoptosis and signaling pathway studies. CAS 51-85-4; formula C4H12N2S2; MW 152.28 g/mol.
  • Supplied as a ready-to-use 10 mM stock solution in DMSO.
  • Convenient 1 mL aliquot for small-scale assays.
  • Inhibits transglutaminase and caspase-3 in biochemical assays.
  • Recommended storage: pure form -20°C; in solvent -80°C or -20°C.
  • Datasheet includes preparation guidance for stock solutions.