Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cystamine dihydrochloride

Catalog No. C121509100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C121509100G 100 g
C12150925G 25 g
C121509500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C121509100G Supplier Sigma Aldrich Supplier No. C121509100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Cystamine dihydrochloride

Specifications

CAS 56-17-7
Melting Point 217°C to 220°C (dec.) (lit.)
Molecular Formula C4H12N2S2 -+ 2 HCl
Linear Formula NH2CH2CH2SSCH2CH2NH2 -+ 2 HCl
MDL Number MFCD00012905
Quantity 100 g
Synonym 2,2 -Diaminodiethyl disulfide dihydrochloride; 2,2 -Dithiobis(ethylamine) dihydrochloride; Decarboxycystine dihydrochloride
Molecular Weight (g/mol) 225.2
Percent Purity 96%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.