Learn More
Apexbio Technology LLC Cystamine dihydrochloride 56-17-7 50mg
SDP
Catalog No. 502240414
Encompass_Preferred
Cystamine dihydrochloride (CAS 56-17-7) is a disulfide-linked derivative of cysteamine that functions as an inhibitor of transglutaminase (Tgase) Additionally it exhibits inhibitory activity against the cysteine protease caspase-3 with a reported IC of approximately 23 6 M Cystamine dihydrochloride is orally bioactive and is commonly utilized in studies investigating the molecular mechanisms and therapeutic strategies for neurodegenerative diseases particularly Huntington s disease Its dual modulatory roles make it valuable for modeling cellular processes relevant to disease pathology
This item is not returnable. View return policy
Catalog No. 50-224-0414 Supplier Apexbio Technology LLC Supplier No. B578550
Cystamine dihydrochloride (CAS 56-17-7) is a disulfide-linked derivative of cysteamine that functions as an inhibitor of transglutaminase (Tgase) Additionally it exhibits inhibitory activity against the cysteine protease caspase-3 with a reported IC of approximately 23 6 M Cystamine dihydrochloride is orally bioactive and is commonly utilized in studies investigating the molecular mechanisms and therapeutic strategies for neurodegenerative diseases particularly Huntington s disease Its dual modulatory roles make it valuable for modeling cellular processes relevant to disease pathology