Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cystamine dihydrochloride | 56-17-7 | MFCD00012905 | ≥98.0% | 225.20 g/mol | C4H14Cl2N2S2 | 10 G
SDP

Catalog No. 5000300465
Encompass_Preferred

Cystamine dihydrochloride is the disulfide form of cysteamine used as a research reagent. It functions as a transglutaminase (Tgase) inhibitor and has reported caspase-3 inhibitory activity (IC50 23.6 μM). Supplied as a solid, it is used in biochemical assays and neurodegeneration studies, including Huntington's disease research.

  • Molecular weight 225.20 g/mol.
  • Chemical formula C4H14Cl2N2S2.
  • High purity (≥98.0% by NMR).
  • Available in multiple solid pack sizes including 10 g.
  • Suitable for Tgase inhibition and caspase activity assays.
  • For laboratory research use; follow appropriate safety procedures.

Catalog No. 50-003-00465 Supplier Medchemexpress LLC Supplier No. HYW02005010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cystamine dihydrochloride is the disulfide form of cysteamine used as a research reagent. It functions as a transglutaminase (Tgase) inhibitor and has reported caspase-3 inhibitory activity (IC50 23.6 μM). Supplied as a solid, it is used in biochemical assays and neurodegeneration studies, including Huntington's disease research.

  • Molecular weight 225.20 g/mol.
  • Chemical formula C4H14Cl2N2S2.
  • High purity (≥98.0% by NMR).
  • Available in multiple solid pack sizes including 10 g.
  • Suitable for Tgase inhibition and caspase activity assays.
  • For laboratory research use; follow appropriate safety procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.