Learn More
Medchemexpress LLC Cystamine dihydrochloride | 56-17-7 | MFCD00012905 | 99.9% | 225.2 g/mol | C4H14Cl2N2S2 | 1 ML
SDP
Catalog No. 5000300934
Encompass_Preferred
Cystamine dihydrochloride is the dihydrochloride salt of cystamine (the disulfide form of cysteamine), used as a transglutaminase inhibitor and reagent in studies of apoptosis, oxidative stress, and neurodegeneration. Supplied as a ready 10 mM solution in DMSO (1 mL), it is intended for research use with defined storage recommendations.
  • Prepared 10 mM solution in DMSO for convenient use.
  • High purity (99.9%) suitable for biochemical assays.
  • Solid form stable under refrigerated, sealed storage.
  • In-solution storage guidance supports short- and long-term stability.
  • Useful transglutaminase inhibitor for cellular and molecular studies.
Catalog No. 50-003-00934 Supplier Medchemexpress LLC Supplier No. HYW02005010MM/1ML
Cystamine dihydrochloride is the dihydrochloride salt of cystamine (the disulfide form of cysteamine), used as a transglutaminase inhibitor and reagent in studies of apoptosis, oxidative stress, and neurodegeneration. Supplied as a ready 10 mM solution in DMSO (1 mL), it is intended for research use with defined storage recommendations.
  • Prepared 10 mM solution in DMSO for convenient use.
  • High purity (99.9%) suitable for biochemical assays.
  • Solid form stable under refrigerated, sealed storage.
  • In-solution storage guidance supports short- and long-term stability.
  • Useful transglutaminase inhibitor for cellular and molecular studies.