Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cystamine dihydrochloride | 56-17-7 | MFCD00012905 | 99.9% | 225.20 g·mol⁻1 | C4H14Cl2N2S2 | 25 G
SDP

Supplier:  Medchemexpress LLC 25GHYW020050

Encompass_Preferred

Cystamine dihydrochloride is the dihydrochloride salt of cystamine (the disulfide form of cysteamine). It is used as a transglutaminase (Tgase) inhibitor and in research on neurodegenerative disorders; supplied as a white to off-white solid with solubility and storage guidance for laboratory applications.

  • High purity suitable for research applications (approx. 99.9%).
  • Soluble in DMSO and water at high concentrations; protocols provided for in vivo formulation.
  • White to off-white solid with clear appearance and handling guidance.
  • Molecular weight 225.20 g·mol⁻1; formula C4H14Cl2N2S2.
  • Available in multiple pack sizes for laboratory use with sealed storage recommendations.

Catalog No. 50-003-02847


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.