Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cystamine dihydrochloride | 56-17-7 | MFCD00012905 | 99.9% | 225.20 g/mol | C4H14Cl2N2S2 | 100 G
SDP

Catalog No. 5000299481
Encompass_Preferred

Cystamine dihydrochloride is the dihydrochloride salt of cystamine (the disulfide form of cysteamine) used as a transglutaminase (Tgase) inhibitor in biochemical and in vivo studies. The material is supplied as a high-purity solid with documented solubility in DMSO and water and supporting analytical documentation for research applications.

  • Transglutaminase inhibitor for biochemical assays.
  • High purity suitable for research applications.
  • Solid material available in multiple pack sizes for convenience.
  • Soluble in DMSO and water with sonication.
  • Analytical documentation available (COA, H NMR, HPLC).
  • Formulation protocols documented for in vivo use.

Catalog No. 50-002-99481 Supplier Medchemexpress LLC Supplier No. HYW020050100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cystamine dihydrochloride is the dihydrochloride salt of cystamine (the disulfide form of cysteamine) used as a transglutaminase (Tgase) inhibitor in biochemical and in vivo studies. The material is supplied as a high-purity solid with documented solubility in DMSO and water and supporting analytical documentation for research applications.

  • Transglutaminase inhibitor for biochemical assays.
  • High purity suitable for research applications.
  • Solid material available in multiple pack sizes for convenience.
  • Soluble in DMSO and water with sonication.
  • Analytical documentation available (COA, H NMR, HPLC).
  • Formulation protocols documented for in vivo use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.