Learn More
Medchemexpress LLC Cystamine dihydrochloride | 56-17-7 | MFCD00012905 | 99.9% | 225.20 g·mol⁻¹ | C4H14Cl2N2S2 | 50 G
SDP
Catalog No. 5000300880
Encompass_Preferred
Cystamine dihydrochloride is the dihydrochloride salt of cystamine, a disulfide-containing small molecule used in biochemical research. It is commonly used as a transglutaminase inhibitor and as a source of cysteamine for biochemical studies. The compound is supplied as a white to off-white solid with high purity suitable for laboratory research.
Catalog No. 50-003-00880 Supplier Medchemexpress LLC Supplier No. HYW02005050G
Cystamine dihydrochloride is the dihydrochloride salt of cystamine, a disulfide-containing small molecule used in biochemical research. It is commonly used as a transglutaminase inhibitor and as a source of cysteamine for biochemical studies. The compound is supplied as a white to off-white solid with high purity suitable for laboratory research.