Learn More
Cayman Chemical Cystathionine-d4, Purity: ≥99% deuterated forms (d1-d4), 1 mg
SDP
Catalog No. 502913310
Encompass_Preferred
An internal standard for the quantification of cystathionine by GC- or LC-MS
Catalog No. 50-291-3310 Supplier Cayman Chemical Supplier No. 366101 MG
An internal standard for the quantification of cystathionine by GC- or LC-MS