Learn More
TARGETMOL CHEMICALS INC CYSTATHIONINE-GAMMA-LYAS 5MG
SDP
Catalog No. 502579372
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cystathionine-(gamma)-lyase-IN-1 (SHIP-2a) is a selective inhibitor of cystathionine (gamma)-lyase (CSE) enzyme (IC50= 6.3 uM). purity: 99%
Catalog No. 50-257-9372 Supplier TARGETMOL CHEMICALS INC Supplier No. T643545MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Cystathionine-(gamma)-lyase-IN-1 (SHIP-2a) is a selective inhibitor of cystathionine (gamma)-lyase (CSE) enzyme (IC50= 6.3 uM). purity: 99%