Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Randox Cystatin C High Control, 3 x 2 mL, liquid, ready-to-use, high control level, immunoturbidimetric assays, 100% human serum
SDP

Catalog No. NC1555330
Encompass_Preferred

The Randox Acusera Cystatin C High Control is a liquid quality control material designed for the routine monitoring of immunoturbidimetric Cystatin C assays. This ready-to-use, third-party control minimizes waste and reduces costs with its long open vial stability. Formulated with 100% human serum, it delivers target values compatible with automated analyzers in clinical laboratories.

  • Formulated with 100% human serum
  • High control level (Level 3)
  • Open vial stability of 30 days at 2°C - 8°C
  • Intended for in vitro diagnostic use
  • Compatible with automated analyzers
  • Provides assayed target values
  • Volume is 3 x 2 mL
  • Stable to expiry date at 2°C - 8°C when unopened
  • Designed for monitoring performance of immunoturbidimetric assays
  • Third-party quality control

This item is not returnable. View return policy
Catalog No. NC1555330 Supplier Randox Supplier No. CYS5020
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Randox Acusera Cystatin C High Control is a liquid quality control material designed for the routine monitoring of immunoturbidimetric Cystatin C assays. This ready-to-use, third-party control minimizes waste and reduces costs with its long open vial stability. Formulated with 100% human serum, it delivers target values compatible with automated analyzers in clinical laboratories.

  • Formulated with 100% human serum
  • High control level (Level 3)
  • Open vial stability of 30 days at 2°C - 8°C
  • Intended for in vitro diagnostic use
  • Compatible with automated analyzers
  • Provides assayed target values
  • Volume is 3 x 2 mL
  • Stable to expiry date at 2°C - 8°C when unopened
  • Designed for monitoring performance of immunoturbidimetric assays
  • Third-party quality control

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.