Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cystatin C ELISA Kit

Catalog No. BMS2279
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BMS2279 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BMS2279 Supplier Invitrogen™ Supplier No. BMS2279
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

The Human Cystatin C Platinum ELISA is an enzyme-linked immunosorbent assay for the quantitative detection of human Cystatin C cell culture supernatant, serum, and plasma (EDTA, citrate, heparin) have been tested with this assay.

Cystatin C, a member of the type 2 cystatin superfamily, is a potent inhibitor of cysteine proteinases, both endogenous (lysosomal cathepsins), and pathogenic. Cystatin C is a 13 kDa secreted protein which is found in high concentrations in biological fluids and is thought to be expressed by all human nucleated cells. Changes in Cystatin C have been documented in a number of cardiovascular diseases, including atherosclerosis, amyloid angiopathy and myocardial infarction. Its low molecular weight also means that Cystatin C can be used as a marker of renal failure.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01034
Assay Range 46.7-3000 pg/mL
Assay Sensitivity 6.9 pg/mL
Conjugate Unlabeled
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Functional biomarker
Sample Type Cell Lysates, Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227AD8; ARMD11; bA218C14.4 (cystatin C); CST3; CYSC; cystatin; cystatin 3; cystatin C; cystatin C (amyloid angiopathy and cerebral hemorrhage); Cystatin C (cysteine proteinase inhibitor); Cystatin3; cystatin-3; CystatinC; Cystatin-C; epididymis secretory protein Li 2; fb51d07; Gamma-trace; HEL-S-2; MGC117328; neuroendocrine basic polypeptide; OTTHUMP00000030440; OTTHUMP00000164181; OTTHUMP00000164182; OTTMUSP00000016736; Post-gamma-globulin; RP23-250M11.9; wu:fb24g06; wu:fb51d07; wu:fc55f03; zgc:136227;
Gene ID (Entrez) 1471
Gene Symbol Cst3
Interassay CV 2.9%
Intraassay CV 6.1%
Kit Contents Pre-coated 96 well plate, Standard (2 vials), Assay Buffer concentrate, HRP-conjugated Antibody, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers, Standard (2 vials), Assay Buffer concentrate, HRP-conjugated Antibody, Wash Buffer, Substrate Solution, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases CST3, ARMD11, HEL-S-2, cystatin C
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Cell Lysate 10 μL, Plasma 10 μL, Serum 10 μL, Supernatant 10 μL
Shipping Condition Wet Ice
Storage Requirements 2-8°C
Target Cystatin C
Test Time 1 hr
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.