Learn More
Randox Cystatin C Control Level 2, 3 x 2 mL
SDP
Catalog No. NC1555329
Encompass_Preferred
The Randox Acusera Cystatin C quality control is designed to monitor the performance of immunoturbidimetric Cystatin C assays. An open vial stability of 30 days at 2°C – 8°C significantly minimises waste and helps to reduce costs.
This item is not returnable. View return policy
Catalog No. NC1555329 Supplier Randox Supplier No. CYS5019
The Randox Acusera Cystatin C quality control is designed to monitor the performance of immunoturbidimetric Cystatin C assays. An open vial stability of 30 days at 2°C – 8°C significantly minimises waste and helps to reduce costs.