Learn More
Medchemexpress LLC Cystatin SN/CST1 Hu 5ug | 5ug
SDP
Supplier:  Medchemexpress LLC HYP78675UG
Encompass_Preferred
Cystatin SN (CST1) is a component of human saliva that is immunologically similar to cystatin S but exhibits distinct specificity due to sequence variation Cystatin SN has an isoelectric point of 7 5 and is a potent inhibitor of papain and dipeptidyl peptidase I superior to Cystatin S in these activities Cystatin SN/CST1 Protein Human (HEK293 His) is the recombinant human-derived Cystatin SN/CST1 protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-94484