Learn More
ACROBiosystems Recombinant Protein;Human Cystatin S / CST4 Protein;HEK293;1MG;CS4,CST4,Cystatin-4,Cystatin-S,Cystatin-S5,CST-4,Cystatin-SA-III,Cystatin S
Supplier:  ACROBiosystems CS4H52231MG
Encompass_Preferred
Recombinant Protein;Human Cystatin S / CST4 Protein;1MG;Human Cystatin S, His Tag (CS4-H5223) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 141 (Accession # AAH65714).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15 kDa. The protein migrates as 15 kDa under reducing (R) condition (SDS-PAGE).
Catalog No. 50-165-8139