Learn More
Cayman Chemical CysteamIn-d4hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 332975 MG
Encompass_Preferred
An internal standard for the quantification of cysteamine by GC- or LC-MS
Catalog No. 50-295-0592