Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Cysteamine

Catalog No. M97685G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M97685G 5 g
M976825G 25 g
M976850G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M97685G Supplier Sigma Aldrich Supplier No. M97685G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Cysteamine is an amino thiol drug mainly used in the treatment of cystinosis
  • It is an antioxidant with chemo-sensitizing and radioprotective properties.tandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf
  • M9768.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database
  • To learn more, visit sigma.com/metlin

Specifications

CAS 60-23-1
Molecular Formula C2H7NS
Linear Formula NH2CH2CH2SH
MDL Number MFCD00008196
Quantity 5 g
Synonym beta-MEA; beta-Mercaptoethylamine; 2-Aminoethanethiol; 2-Mercaptoethylamine; Decarboxycysteine; MEA; Thioethanolamine
Molecular Weight (g/mol) 77.15
Percent Purity ∼95%
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.