Learn More
Selleck Chemical LLC Cysteamine bitartrate
SDP
Supplier:  Selleck Chemical LLC E01785MG
Encompass_Preferred
Cysteamine bitartrate (Mercaptamine bitartrate) is an FDA-approved cystine-depleting aminothiol agent for the treatment of nephropathic cystinosis to decrease accumulation of lysosomal cystine by engaging in disulfide exchange In addition cysteamine bitartrate has antioxidant properties
Catalog No. 50-313-7588