Learn More
Medchemexpress LLC Cysteamine-d4 hydrochloride | 1219805-04-5 | 95.1% | C2H4D4ClNS | 1 MG
SDP
Catalog No. 5000432712
Encompass_Preferred
Cysteamine-d4 hydrochloride is a deuterium-labeled form of Cysteamine hydrochloride. Cysteamine hydrochloride is an orally active agent used in the treatment of nephropathic cystinosis and as an antioxidant. This compound serves as a tracer and an internal standard for quantitative analysis, leveraging the potential of deuteration to influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
  • Orally active for nephropathic cystinosis treatment
  • Acts as an antioxidant
Catalog No. 50-004-32712 Supplier Medchemexpress LLC Supplier No. HY77591S1MG
Cysteamine-d4 hydrochloride is a deuterium-labeled form of Cysteamine hydrochloride. Cysteamine hydrochloride is an orally active agent used in the treatment of nephropathic cystinosis and as an antioxidant. This compound serves as a tracer and an internal standard for quantitative analysis, leveraging the potential of deuteration to influence pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs
  • Orally active for nephropathic cystinosis treatment
  • Acts as an antioxidant