Learn More
Medchemexpress LLC Cysteamine-d4 (hydrochloride) | 1219805-04-5 | 95.1% | C2H4D4ClNS | 5 MG
SDP
Catalog No. 5000435088
Encompass_Preferred
Cysteamine-d4 (hydrochloride) is the deuterium labeled Cysteamine hydrochloride. Cysteamine hydrochloride (2-Aminoethanethiol hydrochloride) is an orally active agent for the treatment of nephropathic cystinosis and an antioxidant.
  • Can be used as a tracer for quantitation.
  • Can be used as an internal standard for quantitative analysis.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
  • For research use only.
Catalog No. 50-004-35088 Supplier Medchemexpress LLC Supplier No. HY77591S5MG
Cysteamine-d4 (hydrochloride) is the deuterium labeled Cysteamine hydrochloride. Cysteamine hydrochloride (2-Aminoethanethiol hydrochloride) is an orally active agent for the treatment of nephropathic cystinosis and an antioxidant.
  • Can be used as a tracer for quantitation.
  • Can be used as an internal standard for quantitative analysis.
  • Deuteration may affect pharmacokinetic and metabolic profiles of drugs.
  • For research use only.