Learn More
Medchemexpress LLC Cysteamine-d4 hydrochloride | 1219805-04-5 | MFCD08704995 | 99.0% | 117.63 | C2H4D4ClNS | 10 MG
SDP
Catalog No. 5000299104
Encompass_Preferred
Cysteamine-d4 hydrochloride is the deuterium-labeled hydrochloride salt of cysteamine provided as a research-grade isotopically labeled reagent for analytical and biochemical studies. It is designed for use where deuterium labeling improves detection, quantification, or tracing in laboratory methods.
  • Deuterium-labeled analog for isotope tracing and internal standards.
  • High purity suitable for analytical applications.
  • Defined molecular weight and formula for accurate mass calculations.
  • Applicable to mass spectrometry, metabolic profiling, and mechanistic studies.
  • Available in small research pack sizes for method development and validation.
Catalog No. 50-002-99104 Supplier Medchemexpress LLC Supplier No. HY77591S10MG
Cysteamine-d4 hydrochloride is the deuterium-labeled hydrochloride salt of cysteamine provided as a research-grade isotopically labeled reagent for analytical and biochemical studies. It is designed for use where deuterium labeling improves detection, quantification, or tracing in laboratory methods.
  • Deuterium-labeled analog for isotope tracing and internal standards.
  • High purity suitable for analytical applications.
  • Defined molecular weight and formula for accurate mass calculations.
  • Applicable to mass spectrometry, metabolic profiling, and mechanistic studies.
  • Available in small research pack sizes for method development and validation.