Learn More
Apexbio Technology LLC Cysteamine HCl 156-57-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094678
Encompass_Preferred
Cysteamine HCl (156-57-0) is a small-molecule modulator designed to facilitate cystine export from lysosomes by lowering intracellular cystine levels Cysteamine HCl exerts its biological activity primarily through cleaving cystine disulfide bonds and forming mixed disulfides enabling cystine transport out of lysosomes In in vitro assays Cysteamine HCl demonstrates antioxidant activity with reported IC50 values typically ranging between 30 80 M depending on cell type and assay conditions Based on these pharmacological properties Cysteamine HCl holds research potential in studies of cystinosis lysosomal storage disorders oxidative stress-related models and cystine metabolic pathway analysis
Catalog No. 50-000-94678 Supplier Apexbio Technology LLC Supplier No. B17195.1
Cysteamine HCl (156-57-0) is a small-molecule modulator designed to facilitate cystine export from lysosomes by lowering intracellular cystine levels Cysteamine HCl exerts its biological activity primarily through cleaving cystine disulfide bonds and forming mixed disulfides enabling cystine transport out of lysosomes In in vitro assays Cysteamine HCl demonstrates antioxidant activity with reported IC50 values typically ranging between 30 80 M depending on cell type and assay conditions Based on these pharmacological properties Cysteamine HCl holds research potential in studies of cystinosis lysosomal storage disorders oxidative stress-related models and cystine metabolic pathway analysis