Learn More
Medchemexpress LLC Cysteamine (hydrochloride) | 156-57-0 | 99.3% | C2H8ClNS | 10 G
SDP
Catalog No. 5000434941
Encompass_Preferred
Cysteamine (hydrochloride) is an orally active agent used for the treatment of nephropathic cystinosis and as an antioxidant. This compound is intended for research use only.
  • Increases intracellular glutathione levels in cystinotic cells, restoring the altered redox state.
  • Counteracts increased rates of apoptosis in cystinotic cells.
  • Exhibits antioxidant properties by increasing glutathione production.
  • Acts as an excellent scavenger of OH and HOCl, and reacts with H2O2.
  • Increases the production of several heat shock proteins (HSP).
  • Exerts a dose-dependent effect on doxorubicin-induced cancer cell death.
Catalog No. 50-004-34941 Supplier Medchemexpress LLC Supplier No. HY7759110G
Cysteamine (hydrochloride) is an orally active agent used for the treatment of nephropathic cystinosis and as an antioxidant. This compound is intended for research use only.
  • Increases intracellular glutathione levels in cystinotic cells, restoring the altered redox state.
  • Counteracts increased rates of apoptosis in cystinotic cells.
  • Exhibits antioxidant properties by increasing glutathione production.
  • Acts as an excellent scavenger of OH and HOCl, and reacts with H2O2.
  • Increases the production of several heat shock proteins (HSP).
  • Exerts a dose-dependent effect on doxorubicin-induced cancer cell death.