Learn More
Medchemexpress LLC Cysteamine (hydrochloride) | 156-57-0 | 99.3% | C2H8ClNS | 25 G
SDP
Catalog No. 5000358656
Encompass_Preferred
Cysteamine hydrochloride is an orally active agent for treating nephropathic cystinosis and functions as an antioxidant. It elevates intracellular glutathione in cystinotic cells, restoring redox balance and mitigating apoptosis. The compound also exhibits antioxidant properties by scavenging reactive oxygen species and boosting glutathione production, and it enhances the production of heat shock proteins.
  • Treats nephropathic cystinosis and acts as an antioxidant.
  • Increases intracellular glutathione levels in cystinotic cells.
  • Counteracts increased apoptosis in cystinotic cells.
  • Scavenges OH, HOCl, and reacts with H2O2.
  • Enhances production of heat shock proteins (HSP).
  • Exerts dose-dependent effect on doxorubicin-induced cancer cell death.
Catalog No. 50-003-58656 Supplier Medchemexpress LLC Supplier No. 25GHY77591
Cysteamine hydrochloride is an orally active agent for treating nephropathic cystinosis and functions as an antioxidant. It elevates intracellular glutathione in cystinotic cells, restoring redox balance and mitigating apoptosis. The compound also exhibits antioxidant properties by scavenging reactive oxygen species and boosting glutathione production, and it enhances the production of heat shock proteins.
  • Treats nephropathic cystinosis and acts as an antioxidant.
  • Increases intracellular glutathione levels in cystinotic cells.
  • Counteracts increased apoptosis in cystinotic cells.
  • Scavenges OH, HOCl, and reacts with H2O2.
  • Enhances production of heat shock proteins (HSP).
  • Exerts dose-dependent effect on doxorubicin-induced cancer cell death.