Learn More
Medchemexpress LLC Cysteamine (hydrochloride) | 156-57-0 | 99.3% | C2H8ClNS | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY77591R
Encompass_Preferred
Cysteamine hydrochloride is an orally active agent used for the treatment of nephropathic cystinosis and acts as an antioxidant. It is intended for research use only.
  • Increases intracellular glutathione levels in cystinotic cells
  • Counteracts increased rates of apoptosis in cystinotic cells
  • Exhibits antioxidant properties
  • Acts as an excellent scavenger of OH and HOCl
  • Increases production of several heat shock proteins
Catalog No. 50-003-59224