Learn More
Medchemexpress LLC Cysteamine hydrochloride | 156-57-0 | 99.6% | C₂H₈ClNS | 250MG
SDP
Catalog No. 5000358486
Encompass_Preferred
This product is an orally active agent used for the treatment of nephropathic cystinosis. It also acts as an antioxidant. It increases intracellular glutathione levels in certain cells, thereby restoring the altered redox state and counteracting increased rates of apoptosis in these cells. It possesses antioxidant properties by increasing glutathione production and is an effective scavenger of certain reactive oxygen species. Additionally, it increases the production of several heat shock proteins and can influence the effectiveness of certain cancer cell treatments.
  • Orally active.
  • Treats a specific kidney disorder.
  • Acts as an antioxidant.
  • Increases intracellular glutathione levels.
  • Restores altered cellular redox state.
  • Counteracts increased apoptosis in certain cells.
  • Scavenges reactive oxygen species.
  • Boosts production of heat shock proteins.
Catalog No. 50-003-58486 Supplier Medchemexpress LLC Supplier No. 250MGHY77591R
This product is an orally active agent used for the treatment of nephropathic cystinosis. It also acts as an antioxidant. It increases intracellular glutathione levels in certain cells, thereby restoring the altered redox state and counteracting increased rates of apoptosis in these cells. It possesses antioxidant properties by increasing glutathione production and is an effective scavenger of certain reactive oxygen species. Additionally, it increases the production of several heat shock proteins and can influence the effectiveness of certain cancer cell treatments.
  • Orally active.
  • Treats a specific kidney disorder.
  • Acts as an antioxidant.
  • Increases intracellular glutathione levels.
  • Restores altered cellular redox state.
  • Counteracts increased apoptosis in certain cells.
  • Scavenges reactive oxygen species.
  • Boosts production of heat shock proteins.