Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteamine hydrochloride, 98.0-101.0% (RT), MilliporeSigma™ Supelco™

Catalog No. 111019903 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9903 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9903 Supplier MilliporeSigma Supelco Supplier No. 49705100MG
Add to Cart
Add to Cart

Specifications

CAS 156-57-0
Linear Formula HSCH2 CH2 NH2 · HCl
MDL Number MFCD00012904
Melting Point 67°C to 71°C
Synonym beta-Mercapto­ethylamine hydrochloride; 2-Amino­ethane­thiol hydrochloride; 2-Mercapto­ethyl­amine hydrochloride; Decarboxy­cysteine hydrochloride; Thio­ethanol­amine hydrochloride
Type Analytical Standard
Quantity 100 mg
Formula Weight 113.61
Percent Purity 98.0 to 101.0% (RT)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.