Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cysteamine hydrochloride (Standard) | 156-57-0 | 99.58% | C2H8ClNS | 100 MG
SDP

Catalog No. 5000435087
Encompass_Preferred

Cysteamine hydrochloride (Standard) is the analytical standard of Cysteamine hydrochloride, intended for research and analytical applications. It is also known as 2-Aminoethanethiol hydrochloride and 2-Mercaptoethylamine hydrochloride. This compound is an orally active agent used in the treatment of nephropathic cystinosis and acts as an antioxidant.

  • Application: commonly used as a reference standard in qualitative, quantitative, and methodological research experiments such as hplc, gc, and ms.
  • Appearance: solid
  • Color: white to off-white
  • Initial source: endogenous metabolite
  • Shipping: room temperature in continental us; may vary elsewhere.

Catalog No. 50-004-35087 Supplier Medchemexpress LLC Supplier No. HY77591R100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cysteamine hydrochloride (Standard) is the analytical standard of Cysteamine hydrochloride, intended for research and analytical applications. It is also known as 2-Aminoethanethiol hydrochloride and 2-Mercaptoethylamine hydrochloride. This compound is an orally active agent used in the treatment of nephropathic cystinosis and acts as an antioxidant.

  • Application: commonly used as a reference standard in qualitative, quantitative, and methodological research experiments such as hplc, gc, and ms.
  • Appearance: solid
  • Color: white to off-white
  • Initial source: endogenous metabolite
  • Shipping: room temperature in continental us; may vary elsewhere.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.