Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cysteamine S-phosphate sodium | 3724-89-8 | MFCD00038474 | 97.0% | 179.11 g/mol | C2H7NNaO3PS | 50 MG
SDP

Catalog No. 5000283563
Encompass_Preferred

Cysteamine S-phosphate sodium is a laboratory research reagent - the sodium salt derivative of cysteamine S-phosphate - used in studies of phosphatase-mediated hydrolysis and cysteamine metabolism. It is supplied as a solid for in vitro and preclinical research applications and is not for therapeutic use.

  • High purity 97.0%.
  • Molecular weight 179.11 g/mol.
  • Molecular formula C2H7NNaO3PS.
  • Available as a 50 mg vial.
  • Appearance white to off-white solid.
  • Store sealed, away from moisture; solution storage: -80°C up to 6 months or -20°C up to 1 month.

Catalog No. 50-002-83563 Supplier Medchemexpress LLC Supplier No. HYW14178150MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cysteamine S-phosphate sodium is a laboratory research reagent - the sodium salt derivative of cysteamine S-phosphate - used in studies of phosphatase-mediated hydrolysis and cysteamine metabolism. It is supplied as a solid for in vitro and preclinical research applications and is not for therapeutic use.

  • High purity 97.0%.
  • Molecular weight 179.11 g/mol.
  • Molecular formula C2H7NNaO3PS.
  • Available as a 50 mg vial.
  • Appearance white to off-white solid.
  • Store sealed, away from moisture; solution storage: -80°C up to 6 months or -20°C up to 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.