Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteamine S-phosphate sodium salt, MilliporeSigma™ Supelco™

Catalog No. 111014071 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4071 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4071 Supplier MilliporeSigma Supelco Supplier No. C839750MG
Add to Cart
Add to Cart

Specifications

Packaging Poly Bottle
CAS 3724-89-8
Physical Form Neat
Linear Formula C2H7 NO3 PSNa
MDL Number MFCD00038474
Synonym S-(2-Aminoethyl) thiophosphate monosodium salt
Type Analytical Standard
Quantity 50 mg
Formula Weight 179.11
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.