Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine Hydrochloride, Monohydrate, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18601961 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-961 25 g Poly Bottle
18-601-962 100 g Poly Bottle
18-601-963 500 g Poly Bottle
18-601-964 1 kg Poly Bottle
18-601-966 50 kg Fiber Drum
18-601-965 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-961 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C147325GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1473, 7048-04-6, C3H7NO2S·HCl·H2O

Spectrum™ Chemical Cysteine Hydrochloride, Monohydrate, USP is a semi-essential amino acid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
IUPAC Name hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
SMILES [H+].O.[Cl-].N[C@@H](CS)C(O)=O

Specifications

CAS Max % 100%
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade USP
Ignition Residue 0.4%
Loss on Drying 8.0-12%
Packaging Poly Bottle
Specific Rotation +5.7° to +6.8°
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.