Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cysteine Hydrochloride, Monohydrate, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18601966 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-966 50 kg Fiber Drum
18-601-961 25 g Poly Bottle
18-601-962 100 g Poly Bottle
18-601-963 500 g Poly Bottle
18-601-964 1 kg Poly Bottle
18-601-965 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-966 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C147350KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

C1473, 7048-04-6, C3H7NO2S·HCl·H2O

Spectrum™ Chemical Cysteine Hydrochloride, Monohydrate, USP is a semi-essential amino acid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4-6-7048
Molecular Formula C3H10ClNO3S
Molecular Weight (g/mol) 175.63
InChI Key QIJRTFXNRTXDIP-JIZZDEOASA-N
IUPAC Name hydrogen (2R)-2-amino-3-sulfanylpropanoic acid hydrate chloride
SMILES [H+].O.[Cl-].N[C@@H](CS)C(O)=O

Specifications

CAS Max % 100%
Quantity 50 kg
Ignition Residue 0.4%
Loss on Drying 8.0-12%
Packaging Fiber Drum
Specific Rotation +5.7° to +6.8°
Percent Purity 98.5 to 101.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.