Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cysteine Protease inhibitor hydrochloride | 2197053-49-7 | 96.0% | 338.79 | 25 MG
SDP

Catalog No. 5000384616
Encompass_Preferred

Cysteine Protease inhibitor hydrochloride is a cysteine protease inhibitor. It can be used in immunoblotting experiments of cells or tissues.

  • Purity: 96.01%
  • Molecular weight: 338.79
  • Formula: C18H15ClN4O
  • Appearance: Solid
  • Color: White to off-white

Catalog No. 50-003-84616 Supplier Medchemexpress LLC Supplier No. 25MGHY17541
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cysteine Protease inhibitor hydrochloride is a cysteine protease inhibitor. It can be used in immunoblotting experiments of cells or tissues.

  • Purity: 96.01%
  • Molecular weight: 338.79
  • Formula: C18H15ClN4O
  • Appearance: Solid
  • Color: White to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.