Learn More
Cayman Chemical Cysteinyl Leukotriene Affinit
SDP
Catalog No. 502942117
Encompass_Preferred
A Cdk2 and Cdk9 inhibitor (Kis 2 and 4 nM respectively) selective for Cdk2 and Cdk9 over Cdk1 Cdk4 and Cdk7 (Kis 80 53 and 70 nM respectively) as well as 10 additional kinases (Kis 1000 nM for all) cytotoxic to A2780 and MES-SA cancer cells (IC50s 41 and 140 nM respectively)
Catalog No. 50-294-2117 Supplier Cayman Chemical Supplier No. 4000681 EA
A Cdk2 and Cdk9 inhibitor (Kis 2 and 4 nM respectively) selective for Cdk2 and Cdk9 over Cdk1 Cdk4 and Cdk7 (Kis 80 53 and 70 nM respectively) as well as 10 additional kinases (Kis 1000 nM for all) cytotoxic to A2780 and MES-SA cancer cells (IC50s 41 and 140 nM respectively)