Learn More
Cayman Chemical Cysteinyl Leukotriene Express
SDP
Catalog No. 502886586
Encompass_Preferred
Cayman’s CysLT Express ELISA Kit is a competitive assay that can be used for quantification of CysLTs in urine, plasma, serum, lavage fluids and aspirates, culture media, and other sample matrices. The assay has a range of 7.8-1,000 pg/ml, an average sensitivity (80% B/B0) of 24 pg/ml, and a lower limit of detection (LLOD) of 6.4 pg/ml. NOTE: This kit measures all CysLTs. For measurement of specific CysLTs, please see Cayman Chemical’s Leukotriene E4 ELISA Kit (Item No. 501060) and Leukotriene C4 ELISA Kit (Item No. 501070).
Catalog No. 50-288-6586 Supplier Cayman Chemical Supplier No. 1000929196 SOLID
Cayman’s CysLT Express ELISA Kit is a competitive assay that can be used for quantification of CysLTs in urine, plasma, serum, lavage fluids and aspirates, culture media, and other sample matrices. The assay has a range of 7.8-1,000 pg/ml, an average sensitivity (80% B/B0) of 24 pg/ml, and a lower limit of detection (LLOD) of 6.4 pg/ml. NOTE: This kit measures all CysLTs. For measurement of specific CysLTs, please see Cayman Chemical’s Leukotriene E4 ELISA Kit (Item No. 501060) and Leukotriene C4 ELISA Kit (Item No. 501070).