Learn More
Enzo Life Sciences Cysteinyl Leukotriene ELISA kit (1x96wells)
SDP
Supplier:  Enzo Life Sciences ADI900070
Encompass_Preferred
Highly sensitive ELISA kit enabling detection of LTC4/D4/E4 in 4 hours. The Cysteinyl Leukotriene EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Safe non-radioactive measurement of LTC4/D4/E4. Ready-to-use liquid color-coded reagents reduce errors. Sensitivity: 26.6 pg/ml (range 78.1 - 2,500 pg/ml). Assay Time: 4 hours. Applications: ELISA, Colorimetric detection. Species Reactivity: Species independent. Contents: GxM IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution
Catalog No. 50-658-128