Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp Cystine Heart Agar 1 KG
SDP

Supplier:  Research Products International Corp C400101000.0

Encompass_Preferred

Supports excellent growth of gram-negative cocci and other pathogenic organisms. With added hemoglobin it is used for cultivation of Francisella tularensis.

Prepare medium by mixing 51 grams of agar medium in 1 Liter of purified water. Mix, heat and autoclave. Cool to 50-60°C. Add 100ml sterile 2% hemoglobin solution. Dispense into sterile petri dish or tube.

Formula per Liter:

Beef Heart Infusion: 10 g
Proteose Peptone: 10 g
Dextrose: 10 g
Sodium Chloride: 5 g
L-Cystine: 1 g
Agar: 15 g

Catalog No. 50-197-7459


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.