Learn More
Bioworld Cystine Heart Agar, 1 kg
Catalog No. 50254266
Encompass_Preferred
Cystine Heart AgarUsed with hemoglobin for cultivating Francisella tularensis and without enrichment for cultivating gram-negative cocci and other microorganismspH: 6.8Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: Plain – light to medium amber agar W/ Hemoglobin – dark brown agarSoluble in boiling waterContains:Beef Heart InfusionPeptoneDextroseSodium ChlorideL-CystineAgarThis product is sold for laboratory research use only.
Catalog No. 50-254-266 Supplier Bioworld Supplier No. 306200022
Cystine Heart AgarUsed with hemoglobin for cultivating Francisella tularensis and without enrichment for cultivating gram-negative cocci and other microorganismspH: 6.8Dehydrated Appearance: beige, homogeneous powderPrepared Appearance: Plain – light to medium amber agar W/ Hemoglobin – dark brown agarSoluble in boiling waterContains:Beef Heart InfusionPeptoneDextroseSodium ChlorideL-CystineAgarThis product is sold for laboratory research use only.