Learn More
Cayman Chemical CystInyl-bIs-glycIn 10mg
SDP
Catalog No. 502990238
Encompass_Preferred
A dipeptide intermediate in the synthesis of oxidized glutathione that is composed of two cysteinylglycine peptides linked by a disulfide bond; formed via nonenzymatic oxidation of cysteinylglycine
Catalog No. 50-299-0238 Supplier Cayman Chemical Supplier No. 3049410 MG
A dipeptide intermediate in the synthesis of oxidized glutathione that is composed of two cysteinylglycine peptides linked by a disulfide bond; formed via nonenzymatic oxidation of cysteinylglycine